missing translation for 'onlineSavingsMsg'
Learn More

Ethanol denatured (Untaxed), UK IDA standard, for license holders in Ireland and United Kingdom only, Honeywell™

Catalog Number 15645600
Click to view available options
Quantity:
2.5 L
25 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5, 67-56-1
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5, 67-56-1
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO
Melting Point -114°C (literature)
CAS Min % ≥50.0000%, ≥3.0000%
CAS Max % ≤100.0000%, <10.0000%
Linear Formula TiCl4
MDL Number MFCD00011267
UN Number UN1987
Beilstein 1718733
Formula Weight 46
Density 0.789 g/mL at 25 °C(lit.)
Boiling Point 78°C to 80°C
Quantity 2.5 L
Flash Point 14°C
Packaging Poly Bottle
Chemical Name or Material Ethanol, Denatured, (UK IDA standard)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethanol; ethyl alcohol, methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'