missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol Absolute, 99.5%, BAKER ANALYZED™, J.T. Baker™
Click to view available options
Quantity:
6 x 500 mL
Packaging:
Glass
Description
BAKER ANALYZED™ reagents are a grade of high quality chemicals for laboratory and specialized industrial use. Every BAKER ANALYZED™ Reagent label provides an Actual Lot Analysis as a guarantee of purity and consistency. Analytical methods used for these determinations are based on standard test procedures described in technical publications.
Specifications
Melting Point | −114.1°C |
Absorbance | Max. 1.00 at 210nm, Max. 0.50 at 220nm, Max. 0.20 at 230nm, Max. 0.05 at 250nm, Max. 0.01 at 270-400nm |
CAS | 64-17-5 |
Color | Colorless |
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.07 |
Percent Purity | Min. 99.5% |
Density | 0.79g/cm3 at 20°C |
Boiling Point | 78.2°C to 78.5°C |
Quantity | 6 x 500 mL |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'