missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ Q-CLASSIC-90 One-Door Yellow Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas.
Brand: Asecos™ 3007204033600
Additional Details : Weight : 314.00000kg
Beschreibung
- Extremely robust design with scratch-proof surface
- Triple-hinged doors
- Safety elements assembled outside the storage compartment for increased protection against corrosion
- Integrated transport base for simple in-house transport
- Cabinet can be transported upright through doors
- Adjustable feet in base allow the safe and fast compensation of minor surface unevenness
- Modern bicolor design - choice of seven different door colors
- Language-free labelling for optimum international usability
- Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
- Locking status indicator (red/green) signals the locking status
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- In accordance with EN 14470-1 and TRGS 510
- Fire resistance of 90 minutes
Spezifikation
615 x 599 x 1953 mm | |
Yellow | |
Safety storage cabinet | |
3 x Shelves 75 kg each; Insert 75 kg; Sump 22 L | |
1 |
313.5 kg | |
3 x shelves; 1 x perforated insert; 1 x bottom collecting sump | |
Storage of flammable hazardous substances | |
Triple-hinged | |
EN 14470-1 |