missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mini Dumont #M5S, Stainless Steel, 8.2cm, 0.09 x 0.05 mm tip
Brand: Dumont 501764
Mini Dumont #M5S, Stainless Steel, 8.2cm, 0.09 x 0.05 mm tip
Code : Z2
Additional Details : Weight : 0.50000kg