missing translation for 'onlineSavingsMsg'
Learn More

15-Crown-5, 98%, Thermo Scientific Chemicals

Catalog Number p-7082032
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
33100-27-5
C10H20O5
220.265
MFCD00005110
VFTFKUDGYRBSAL-UHFFFAOYSA-N
15-crown-5, 15-crown-5 ether, 15-crown 5-ether, 1,4,10,13-pentaoxacyclopentadecane, ccris 3586, 15-crown 5, crown ether 15-crown-5, 5-19-12-00252 beilstein handbook reference, ksc917c1b
36336
CHEBI:32401
1,4,7,10,13-pentaoxacyclopentadecane
C1COCCOCCOCCOCCO1
This item is not returnable. View return policy
33100-27-5
C10H20O5
220.265
MFCD00005110
VFTFKUDGYRBSAL-UHFFFAOYSA-N
15-crown-5, 15-crown-5 ether, 15-crown 5-ether, 1,4,10,13-pentaoxacyclopentadecane, ccris 3586, 15-crown 5, crown ether 15-crown-5, 5-19-12-00252 beilstein handbook reference, ksc917c1b
36336
CHEBI:32401
1,4,7,10,13-pentaoxacyclopentadecane
C1COCCOCCOCCOCCO1

15-Crown-5 is used as an efficient phase transfer catalyst and as a complexing agent. It is used to isolate oxonium ion (H7O3)+ salts especially from a solution of tetrchloroauric acid. It catalyzes the O-alkylation of the sodium salts of carboxylic acids in the penicillin and cephalosporin series, thereby facilitating esterification reaction without usage of acid. It is also used to facilitate the Williamson synthesis of ethers with hindered alcohols and sodium hydride. It is used with lithium aluminum hydride, for performing reduction reactions in hydrocarbon solvents. Further, it is involved in the Horner-Wadsworth-Emmons reaction to prepare stilbenes from aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
15-Crown-5 is used as an efficient phase transfer catalyst and as a complexing agent. It is used to isolate oxonium ion (H7O3)+ salts especially from a solution of tetrchloroauric acid. It catalyzes the O-alkylation of the sodium salts of carboxylic acids in the penicillin and cephalosporin series, thereby facilitating esterification reaction without usage of acid. It is also used to facilitate the Williamson synthesis of ethers with hindered alcohols and sodium hydride. It is used with lithium aluminum hydride, for performing reduction reactions in hydrocarbon solvents. Further, it is involved in the Horner-Wadsworth-Emmons reaction to prepare stilbenes from aldehydes.

Solubility
Miscible with organic solvents.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Density 1.11
Boiling Point 150°C to 155°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.465
Quantity 1 g
Beilstein 1618144
Sensitivity Hygroscopic
Solubility Information Miscible with organic solvents.
Formula Weight 220.27
Percent Purity 98%
Chemical Name or Material 15-Crown-5
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'