missing translation for 'onlineSavingsMsg'
Learn More

Linden Fuse for plugs to BS 1362 13 A 1 inch

Brand:  Linden H-ER-FUSE13

Fuse for plugs to BS 1362 13 A 1 inch

Code : 33

Additional Details : Weight : 1.00000kg

Product Code. 12723646

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.