Learn More
Thermo Scientific™ Hypersep™ Hypercarb™ Fixed Well Plate
Unique material for retention of highly polar compounds.
Brand:  Thermo Scientific™ 60306-502
Code : A0
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 13496976
  • £451.98 / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
Thermo Scientific™ HyperSep™ and Hypercarb™ SPE contains flat,100% porous graphitic carbon (PGC) with selectivity for structurally similar compounds, offering separation of compounds with simple solvents
  • pH stable 0 to 14
  • High batch-to-batch reproducibility
  • Strong retention properties allow use of low bed weights for concentrated extracts
  • Interaction mechanism with polar molecules
Specifications
Specifications
25 mg
40 to 60 μm
HypeSep Hypercarb
1/Ea
Universal Vacuum Manifold
HyperSep
96-well Plate