Learn More
SI Analytics™ Titration Stand with Pump
SI Analytics™ Titration Stand with Pump comes equipped with a reagent bottle, a waste bottle, a moisture bottle, tubes, and a power supply.
Brand:  SI Analytics™ 285220900
Code : NEW
Additional Details : Weight : 3.00000kg
Product Code. 13416778
  • £1355.00 / Each
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • Scope of delivery: Basic unit with 1 DURAN™-reagent bottle TZ 1791, 1 DURAN™-waste bottle TZ 1792, moisture bottle, tubes and screw threads, power supply TZ 1855 (110 to 240 V)
Specifications
Specifications
Titration Stand
Basic unit with 1 DURAN™-reagent bottle TZ 1791, 1 DURAN™-waste bottle TZ 1792, moisture bottle, tubes and screw threads, power supply TZ 1855 (110 to 240 V)