Learn More
Thermo Scientific™ Hypersep™ Hypercarb™ Fixed Well Plate
Unique material for retention of highly polar compounds.
Brand: Thermo Scientific™ 60306-503
Code : A0
Additional Details : Weight : 0.50000kg
Description
Thermo Scientific™ HyperSep™ and Hypercarb™ SPE contains flat,100% porous graphitic carbon (PGC) with selectivity for structurally similar compounds, offering separation of compounds with simple solvents
- pH stable 0 to 14
- High batch-to-batch reproducibility
- Strong retention properties allow use of low bed weights for concentrated extracts
- Interaction mechanism with polar molecules
Specifications
50 mg | |
40 to 60 μm | |
HypeSep Hypercarb | |
1/Ea |
Universal Vacuum Manifold | |
HyperSep | |
96-well Plate |