missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium hydrogen phosphate dihydrate, BAKER ANALYZED™, J.T. Baker™
111.26 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
1 kg
Specifications
CAS | 10028-24-7 |
pH | 9.0 to 9.5 (0.1 M Solution (at 25°C)) |
Quantity | 1 kg |
Assay Percent Range | min. 99% |
Molecular Formula | H5Na2O6P |
MDL Number | MFCD00149182 |
Synonym | Sorensen's salt , di-Sodium hydrogen orthophosphate dihydrate , Sodium phosphate dibasic dihydrate , Sodium monohydrogen phosphate dihydrate |
InChI Key | KDQPSPMLNJTZAL-UHFFFAOYSA-L |
SMILES | O.O.[Na+].[Na+].OP([O-])([O-])=O |
IUPAC Name | disodium dihydrate hydrogen phosphate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'