missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tweezers Dumont #1 straight Inox 0.6mm tip 120mm
Brand: WPI World Precision Instrument 500336
Tweezers Dumont #1 straight Inox 0.6mm tip 120mm
Code : Z2
Additional Details : Weight : 0.10000kg