Learn More
X100 Syringe filter polypropylene polytetrafluoroethylene (PTFE)membrane 0.22µm 13mm (pack of 100)
Brand:  Kinesis 12916-60
X100 Syringe filter polypropylene polytetrafluoroethylene (PTFE)membrane 0.22µm 13mm (pack of 100)
Code : Z4
Additional Details : Weight : 0.25000kg
Product Code. 13003020
  • £157.00 / Pack of 100