missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, BAKER ANALYZED™ LC-MS Reagent, J.T. Baker™
Click to view available options
1 L
Description
- LC-Gradient-Diode Array Detection (AU), test solution is modified with 0.1% (v/v) formic acid:
- at 235 nm max. 0.005
- at 254 nm max. 0.001
- LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), test solution is modified with 0.1% (v/v) formic acid:
- Positive ESI-MS Sensitive Impurities (as Reserpine) max. 50 ng/mL
- Ultraviolet Absorbance (1.00-cm path vs water):
- at 225 nm max. 0.20
- at 254 nm max. 0.02
- at 280 nm max. 0.01
Specifications
Melting Point | -98°C |
CAS | 67-56-1 |
Assay Percent Range | min. 99.8% |
Molecular Formula | CH4O |
MDL Number | MFCD00004595 |
UN Number | 1230 |
Synonym | Hydroxy methane , Methyl alcohol , Carbinol , Wood alcohol |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
SMILES | CO |
IUPAC Name | methanol |
Show More |
Safety and Handling
Product Identifier
- Methanol
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'