missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 12-Tricosanone, 97%

Antiblocking agent

Brand:  Alfa Aesar™ 041444.22

Code : NEW

Additional Details : CAS Number : 540-09-0 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11354168

  • £149.99 / 100g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

12-Tricosanone
338.62
Diundecyl ketone
540-09-0
MFCD00026562

Specifications

12-Tricosanone
540-09-0
97%
MFCD00026562
66° to 68°C
100g
Powder
338.62
Diundecyl ketone
Videos
SDS