missing translation for 'onlineSavingsMsg'
Learn More

12-Crown-4, 98%, Thermo Scientific Chemicals

Catalog Number p-7081780
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
294-93-9
C8H16O4
176.21
MFCD00005103
XQQZRZQVBFHBHL-UHFFFAOYSA-N
9269
CHEBI:32399
1,4,7,10-tetraoxacyclododecane
C1COCCOCCOCCO1
This item is not returnable. View return policy
294-93-9
C8H16O4
176.21
MFCD00005103
XQQZRZQVBFHBHL-UHFFFAOYSA-N
9269
CHEBI:32399
1,4,7,10-tetraoxacyclododecane
C1COCCOCCOCCO1

It serves as an aprotic solvent for alkali metals, the sodium solution is a strong reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It serves as an aprotic ″solvent″ for alkali metals, the sodium solution is a strong reducing agent.

Solubility
Fully miscible in water.

Notes
Light & Air Sensitive & Hygroscopic. Store under inert gas. Keep in a tightly closed container.
TRUSTED_SUSTAINABILITY
Melting Point 17°C
Density 1.108
Boiling Point 68°C to 70°C (0.1 mmHg)
Flash Point >109°C (228°F)
Odor Odorless
Refractive Index 1.462
Quantity 1 g
UN Number UN2810
Beilstein 1363064
Sensitivity Air and light sensitive; Hygroscopic
Synonym 12-crown-4, eoct, 12-crown-4 ether, ethylene oxide cyclic tetramer, ccris 157, unii-k6069p2c2a, 12-crown4-ether, 12-crown 4-ether, 5-19-11-00334 beilstein handbook reference, 3,6,9,12-tetraoxacyclododecan
Solubility Information Fully miscible in water.
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material 12-Crown-4
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'