missing translation for 'onlineSavingsMsg'
Learn More
Learn More
12-Bromododecylphosphonic acid, 95%, Thermo Scientific Chemicals
61.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
Employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as a intermediate for pharmaceutical.
Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
Employed as a intermediate for pharmaceutical.
Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 92°C to 94°C |
Linear Formula | Br(CH2)12P(O)(OH)2 |
Quantity | 250 mg |
Formula Weight | 329.21 |
Percent Purity | 95% |
Chemical Name or Material | 12-Bromododecylphosphonic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'