missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Polystrene Petri Dish

Crystal polystyrene dishes, with optimal flatness.

Brand:  Gosselin BP53-06

230.12 GBP valid until 2024-12-31
Use promo code "23288" to get your promotional price.



Code : A0

Additional Details : Weight : 10.13400kg

 View more versions of this product

Product Code. 11758573

  • £395.00 / Pack of 1620
In Stock
to see stock.
Add to basket

Description

Description

  • Appropriate for aseptic production
  • Sterilized by irradiation
  • Flat bottom
Specifications

Specifications

CE, IVD, EC N°1935/2004, ISO 11137, Aseptic
Polystyrene
Sterile
1
6
55 mm
Natural
Round
Petri Dish
For use in clinical, environment, industries, microbiology
14.2 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers