missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ 11301 Series Non-sterile Cellulose Nitrate (CN) Membrane Filters

Brand:  Sartorius™ 11301--25------N

Code : NEW

Additional Details : Weight : 0.10000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 10067960

Quantity Price
1 £113.78 / Pack of 100
10 £107.87 / Pack of 100
Estimated Shipment: 04-01-2022

Add to basket


Description

Description

Specifications

Specifications

Cellulose Nitrate
Cellulose Nitrate
Membrane filter
White
25 mm
Autoclavable
750mL/min./cm2/bar
100/Pk
8 μm
Non-sterile
Membrane Filter
25 mm
130 μm
0.3bar
For many general laboratory applications where a membrane with a high non-specific adsorption is suitable
100
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • CN membrane, 8 µm, 25 mm, 100 pcs

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P230-Keep wetted with .
  • P243-Take precautionary measures against static discharge.
  • P234-Keep only in original container.
  • P233-Keep container tightly closed.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P372-Explosion risk in case of fire.
  • P375-Fight fire remotely due to the risk of explosion.
  • P370+P380-In case of fire: Evacuate area.
  • P501-Dispose of contents/container to .

SDS
Documents
Certificates