missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11-Hydroxyundecylphosphonic acid, 95%, Thermo Scientific Chemicals
181.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Melting Point | 110°C to 112°C |
Linear Formula | HO(CH2)11P(O)(OH)2 |
Quantity | 1 g |
Formula Weight | 252.29 |
Percent Purity | 95% |
Chemical Name or Material | 11-Hydroxyundecylphosphonic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'