missing translation for 'onlineSavingsMsg'
Learn More

11-Bromo-1-undecanol, 97%, Thermo Scientific Chemicals

Catalog Number p-77638
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1611-56-9
C11H23BrO
251.21
MFCD00004752
XFGANBYCJWQYBI-UHFFFAOYSA-N
11-bromo-1-undecanol, 11-bromoundecanol, 1-undecanol, 11-bromo, 1-bromo-11-hydroxyundecane, undecamethylene bromohydrin, 11-bromoundecyl alcohol, 11-bromo-l-undecanol, 11-bromoundecane-1-ol, 11-bromanylundecan-1-ol, acmc-1br3a
74163
11-bromoundecan-1-ol
OCCCCCCCCCCCBr
This item is not returnable. View return policy
1611-56-9
C11H23BrO
251.21
MFCD00004752
XFGANBYCJWQYBI-UHFFFAOYSA-N
11-bromo-1-undecanol, 11-bromoundecanol, 1-undecanol, 11-bromo, 1-bromo-11-hydroxyundecane, undecamethylene bromohydrin, 11-bromoundecyl alcohol, 11-bromo-l-undecanol, 11-bromoundecane-1-ol, 11-bromanylundecan-1-ol, acmc-1br3a
74163
11-bromoundecan-1-ol
OCCCCCCCCCCCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 42°C to 47°C
Color White to Beige
Boiling Point 165°C to 170°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula Br(CH2)11OH
Quantity 10 g
Beilstein 01, IV, 1837
Formula Weight 251.21
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 11-Bromo-1-undecanol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 11-Bromo-1-undecanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'