missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11-Aminoundecyltriethoxysilane, 90+%, Thermo Scientific Chemicals
84.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
Used as primary and secondary intermediate for pharmaceutical and chemical additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as primary and secondary intermediate for pharmaceutical and chemical additives.
Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
Used as primary and secondary intermediate for pharmaceutical and chemical additives.
Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
Specifications
Boiling Point | 110°C to 115°C (0.02mmHg) |
Quantity | 250 mg |
Linear Formula | H2N(CH2)11Si(OCH2CH3)3 |
Synonym | 11-triethoxysilyl-1-undecanamine, 11-aminoundecyltriethoxysilane, 11-triethoxysilyl undecan-1-amine, 1-undecanamine,11-triethoxysilyl, 11-aminoundecyl triethoxysilane, acmc-20mmty, 1-undecanamine, 11-triethoxysilyl |
Formula Weight | 333.59 |
Percent Purity | 95% |
Sensitivity | Moisture sensitive |
Chemical Name or Material | 11-Aminoundecyltriethoxysilane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'