missing translation for 'onlineSavingsMsg'
Learn More

10GR 2-(4-Nitrophenyl)ethanethioamide, 97%

Brand:  Thermo Scientific Maybridge DP01591EA

10GR 2-(4-Nitrophenyl)ethanethioamide, 97%

Code : 31

Additional Details : CAS Number : 76254-70-1 Weight : 0.06000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Product Code. 10710554

  • £284.00 / 10g
Estimated Shipment: 09-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!