missing translation for 'onlineSavingsMsg'
Learn More

Iodine in aqueous solution, BAKER ANALYZED™, J.T. Baker™

Catalog Number 10627781
Click to view available options
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Laboratory Reagent

Concentration 0.05 mol/L (0.1 N)
CAS 7553-56-2
Quantity 1 L
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II
IUPAC Name diiodine
Molecular Weight (g/mol) 253.81
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'