missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, 96% (v/v), BAKER ANALYZED, J.T. Baker™
254.60 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
5 L
Packaging:
EcoTainer Metal Safety Can
Descripción
- Exceeds the requirements set forth by the American Chemical Society (ACS)
- Label provides an actual Lot Analysis as a guarantee of purity and consistency
- Analytical methods used for these determinations are based on standard test procedures described in technical publications
- Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Especificaciones
Melting Point | -114.5°C |
CAS | 64-17-5 |
Color | Colorless |
Assay Percent Range | 95.1 to 96.9% v/v |
Molecular Formula | C2H6O |
Linear Formula | C2H5OH |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
SMILES | CCO |
IUPAC Name | ethanol |
Mostrar más |
Seguridad y manipulación
Nombre del producto
- ethanol absolute
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'