Learn More
Quickfit™ Screwthread Adapter to Fit Cone
Secure thermometers and tubes to glass containers.  The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
Brand:  Quickfit ST51/13
52.35 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : 01
Additional Details : Weight : 0.02520kg
 View more versions of this product
Product Code. 10187988
  • £118.00 / Pack of 2
In Stock
to see stock.
Add to basket
Description
Description
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
  • With screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer
  • Ideal for securing thermometers, gas inlet tubes, etc
Specifications
Specifications
14/23
Plastic cap, Silicone Rubber ring and PTFE washer
Cone screw thread adapter
Ideal for securing thermometers, gas inlet tubes
Borosilicate Glass
13