missing translation for 'onlineSavingsMsg'
Learn More

Endecotts™ 100mm Diameter Stainless Steel Frame and Wire Mesh Test Sieves

Use these test sieves for all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Frame Test Sieves can be supplied with aperture sizes ranging from 125mm down to 20 microns in full and half height versions.

Brand:  Endecotts™ 100SIW9.50

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12944195

  • £113.00 / Each
Estimated Shipment: 04-07-2024
to see stock.
Add to basket

Description

Description

  • Made with only the highest quality materials, available in diameter sizes of 38, 100, 150, 200, 250, 300, 315, 350, 400 or 450mm as well as in 3, 8, 12 or 18 inches.
  • Precision frame (ensures consistent nestability)
  • Precise aperture (in accordance with ISO 3310-1, ASTM E11 or other specifications)
  • Natural fillet (free flowing of sample)
  • Totally sealed (no crevice to lose material)
  • Evenly tensioned mesh (ensures accurate analysis)
  • Safe edge (big radius makes it comfortable to handle)
  • Serial number (ensures full traceability)
Specifications

Specifications

Stainless Steel
Stainless Steel
1Each
Test Sieve
Stainless Steel
For all types of laboratory sampling and particle size analysis
100 mm
9.5mm
ISO 3310-1 & 2 requirements, ASTM, BSI
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers