missing translation for 'onlineSavingsMsg'
Learn More

Endecotts™ 100mm Diameter Brass Frame With Stainless Steel Wire Mesh Test Sieves

Use these test sieves for all types of laboratory sampling and particle size analysis. Endecotts™ Brass Frame Test Sieves can be supplied with aperture sizes ranging from 125mm down to 20 microns in full and half height versions.

Brand:  Endecotts™ 100BIW.212

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12904255

  • £93.40 / Each


Estimated Shipment
20-06-2018
Add to basket

Description

Description

  • Made with only the highest quality materials, available in diameter sizes of 38, 100, 150, 200, 250, 300, 315, 350, 400 or 450mm as well as in 3, 8, 12 or 18 inches.
  • Precision frame (ensures consistent nestability)
  • Precise aperture (in accordance with ISO 3310-1, ASTM E11 or other specifications)
  • Natural fillet (free flowing of sample)
  • Totally sealed (no crevice to lose material)
  • Evenly tensioned mesh (ensures accurate analysis)
  • Safe edge (big radius makes it comfortable to handle)
  • Serial number (ensures full traceability)
Specifications

Specifications

For all types of laboratory sampling and particle size analysis
Brass
212μm
ISO 3310-1 & 2 requirements, ASTM, BSI
Stainless Steel
1Each
100mm
Test Sieve
Brass
Videos