missing translation for 'onlineSavingsMsg'
Learn More

Endecotts™ 100mm Diameter Brass Frame With Stainless Steel Wire Mesh Test Sieves

Use these test sieves for all types of laboratory sampling and particle size analysis. Endecotts™ Brass Frame Test Sieves can be supplied with aperture sizes ranging from 125mm down to 20 microns in full and half height versions.

Brand:  Endecotts™ 100BIW.500

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12954245

  • £102.58 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

For all types of laboratory sampling and particle size analysis
Brass
500μm
ISO 3310-1 & 2 requirements, ASTM, BSI
Stainless Steel
1Each
100mm
Test Sieve
Brass
Videos