missing translation for 'onlineSavingsMsg'
Learn More

1-Undecanol, 98%, Thermo Scientific Chemicals

Catalog Number p-210512
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
112-42-5
C11H24O
172.31
MFCD00004751
KJIOQYGWTQBHNH-UHFFFAOYSA-N
1-undecanol, undecyl alcohol, undecanol, n-undecanol, n-undecyl alcohol, hendecanoic alcohol, hendecyl alcohol, 1-hendecanol, n-hendecylenic alcohol, tip-nip
8184
CHEBI:87499
undecan-1-ol
CCCCCCCCCCCO
This item is not returnable. View return policy
112-42-5
C11H24O
172.31
MFCD00004751
KJIOQYGWTQBHNH-UHFFFAOYSA-N
1-undecanol, undecyl alcohol, undecanol, n-undecanol, n-undecyl alcohol, hendecanoic alcohol, hendecyl alcohol, 1-hendecanol, n-hendecylenic alcohol, tip-nip
8184
CHEBI:87499
undecan-1-ol
CCCCCCCCCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 11.0°C
Color Colorless to Yellow
Density 0.8310g/mL
Boiling Point 146.0°C (30.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)10OH
Refractive Index 1.4370 to 1.4420
Quantity 100 mL
Beilstein 01, 427
Specific Gravity 0.831
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in 60% alcohol
Formula Weight 172.31
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Undecanol, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Undecanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'