missing translation for 'onlineSavingsMsg'
Learn More

1-Trityl-1H-imidazole-4-carboxaldehyde, 95%, ACROS Organics™

Brand:  Acros Organics 429430050

Code : 29

Additional Details : CAS Number : 33016-47-6 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10628550

  • £74.50 / 5g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

1-Trityl-1H-imidazole-4-carboxaldehyde
94.0
95%
C23H18N2O
MFCD02179554
33016-47-6
100.0
95%
338.41

Specifications

1-Trityl-1H-imidazole-4-carboxaldehyde
94.0
Authentic
95%
C23H18N2O
338.41
White to brown
5g
33016-47-6
100.0
Crystalline Powder
95%
Glass bottle
MFCD02179554
186.0° to 189.0°C
Videos
SDS