Learn More
1-(Trimethylsilyl)imidazole, 97%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
1-(Trimethylsilyl)imidazole acts as a silylating agent for alcohols, carbohydrates and 1,3-dicarbonyl compounds. It is a derivatization reagent used for selective silylation of hydroxyl groups and carboxylic acid without affecting the amino group. It employed as an intermediate for the synthesis of imidazole derivatives. It plays an essential role to synthesize polysubstituted chiral spirotetrahydropyrans. It is also used in the quantification of pintol after derivatization by gas chromatography/ mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-(Trimethylsilyl)imidazole acts as a silylating agent for alcohols, carbohydrates and 1,3-dicarbonyl compounds. It is a derivatization reagent used for selective silylation of hydroxyl groups and carboxylic acid without affecting the amino group. It employed as an intermediate for the synthesis of imidazole derivatives. It plays an essential role to synthesize polysubstituted chiral spirotetrahydropyrans. It is also used in the quantification of pintol after derivatization by gas chromatography/ mass spectrometry.
Solubility
Miscible with most common organic solvents. It decomposes in water.
Notes
Moisture and light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
Specifications
Melting Point | -42°C |
Density | 0.956 |
Boiling Point | 212°C |
Flash Point | 18°C (64°F) |
Odor | Weak |
Refractive Index | 1.4765 |
Quantity | 5 g |
UN Number | UN1993 |
Beilstein | 606148 |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only