missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Trimethylsilyl-2-pyrrolidinone, 96%

Brand:  Alfa Aesar™ L02638.09

Code : NEW

Additional Details : CAS Number : 14468-90-7 Weight : 0.01000kg Transport : UN number : 1993 Chem class : 3 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11367775

  • £28.70 / 10g


EU Stock 3
Add to basket

Description

Description

Applications
1-Trimethylsilyl-2-pyrrolidinone, is used as a fine chemical intermediate.

Solubility
It hydrolyzes with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

1-Trimethylsilyl-2-pyrrolidinone
C7H15NOSi
MFCD00051572
114950
14468-90-7
157.29
UN1993
N-Trimethylsilylbutyrolactam; N-Trimethylsilyl-2-pyrrolidone

Specifications

1-Trimethylsilyl-2-pyrrolidinone
90°C (20mmHg)
14468-90-7
C7H15NOSi
MFCD00051572
114950
46°C (115°F)
10g
96%
157.29
UN1993
N-Trimethylsilylbutyrolactam; N-Trimethylsilyl-2-pyrrolidone
Videos
SDS