missing translation for 'onlineSavingsMsg'
Learn More

1-(trans-Cinnamoyl)imidazole, 98%, Thermo Scientific Chemicals

Catalog Number p-7043363
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1138-15-4
C12H10N2O
198.225
MFCD00005288
XVGXMXZUJNAGFZ-VOTSOKGWSA-N
n-trans-cinnamoylimidazole, trans-cinnamoylimidazole, imidazole, 1-cinnamoyl-, e, imidazole, 1-cinnamoyl-, trans, imidazole, trans, 1-trans-cinnamoyl imidazole, imidazole, e, e-1-1h-imidazol-1-yl-3-phenylprop-2-en-1-one, 1h-imidazole, e, 2e-1-imidazolyl-3-phenylprop-2-en-1-one
5357650
(E)-1-imidazol-1-yl-3-phenylprop-2-en-1-one
C1=CC=C(C=C1)C=CC(=O)N2C=CN=C2
This item is not returnable. View return policy
1138-15-4
C12H10N2O
198.225
MFCD00005288
XVGXMXZUJNAGFZ-VOTSOKGWSA-N
n-trans-cinnamoylimidazole, trans-cinnamoylimidazole, imidazole, 1-cinnamoyl-, e, imidazole, 1-cinnamoyl-, trans, imidazole, trans, 1-trans-cinnamoyl imidazole, imidazole, e, e-1-1h-imidazol-1-yl-3-phenylprop-2-en-1-one, 1h-imidazole, e, 2e-1-imidazolyl-3-phenylprop-2-en-1-one
5357650
(E)-1-imidazol-1-yl-3-phenylprop-2-en-1-one
C1=CC=C(C=C1)C=CC(=O)N2C=CN=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 128°C to 132°C
Odor Odorless
Quantity 5 g
Beilstein 608846
Sensitivity Moisture sensitive
Formula Weight 198.23
Percent Purity 98%
Chemical Name or Material 1-(trans-Cinnamoyl)imidazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'