missing translation for 'onlineSavingsMsg'
Learn More

[1-(Thien-2-ylmethyl)piperid-4-yl]methylamine, 97%, Maybridge

Brand:  Maybridge CC66713DA

Code : 31

Additional Details : CAS Number : 883541-34-2 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10072203

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

[1-(Thien-2-ylmethyl)piperid-4-yl]methylamine
94.5
95%
C11H18N2S
MFCD06801314
883541-34-2
100.0
95%
210.34

Specifications

[1-(Thien-2-ylmethyl)piperid-4-yl]methylamine
883541-34-2
100.0
95%
C11H18N2S
MFCD06801314
1g
94.5
Amber Glass Bottle
95%
210.34
Videos
SDS