missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecene, 94%, Thermo Scientific Chemicals

Catalog Number p-7021577
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
1120-36-1
C14H28
196.378
MFCD00008981
HFDVRLIODXPAHB-UHFFFAOYSA-N
1-tetradecene, n-tetradec-1-ene, tetradecene, 1-tetradecylene, alpha-tetradecene, unii-fw23481s7s, ccris 3785, alkenes, c10-16 .alpha., a-tetradecene, tetradecene-1
14260
CHEBI:77505
tetradec-1-ene
CCCCCCCCCCCCC=C
This item is not returnable. View return policy
1120-36-1
C14H28
196.378
MFCD00008981
HFDVRLIODXPAHB-UHFFFAOYSA-N
1-tetradecene, n-tetradec-1-ene, tetradecene, 1-tetradecylene, alpha-tetradecene, unii-fw23481s7s, ccris 3785, alkenes, c10-16 .alpha., a-tetradecene, tetradecene-1
14260
CHEBI:77505
tetradec-1-ene
CCCCCCCCCCCCC=C

1-Tetradecene used in the production of linear plasticizers, oxo-alcohols, motor fuels, lubricants, automotive additives, biodegradable surfactants, paper size, and in a wide range of specialty applications in the production of mercaptans, flavors and fragrances, alkyl metals, halides, alkyl silanes.. It is also used as a co monomers in polymer such as low density polyethylenes for the properties of lighter, thinner, better flexibility and more tearing resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecene used in the production of linear plasticizers, oxo-alcohols, motor fuels, lubricants, automotive additives, biodegradable surfactants, paper size, and in a wide range of specialty applications in the production of mercaptans, flavors and fragrances, alkyl metals, halides, alkyl silanes.. It is also used as a co monomers in polymer such as low density polyethylenes for the properties of lighter, thinner, better flexibility and more tearing resistance.

Solubility
Soluble in hydrocarbon solvents; insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point -12°C
Density 0.775
Boiling Point 250°C to 252°C
Flash Point 115°C (239°F)
Assay Percent Range 94%
Odor Mild
Linear Formula CH3(CH2)11CH=CH2
Refractive Index 1.436
Quantity 100 mL
Beilstein 1701156
Solubility Information Soluble in hydrocarbon solvents; insoluble in water.
Formula Weight 196.38
Percent Purity 94%
Chemical Name or Material 1-Tetradecene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'