missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecanol, 99%, Thermo Scientific Chemicals

Catalog Number 10503551
14.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
112-72-1
C14H30O
214.39
MFCD00004757
HLZKNKRTKFSKGZ-UHFFFAOYSA-N
1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks
8209
CHEBI:77417
tetradecan-1-ol
CCCCCCCCCCCCCCO
This item is not returnable. View return policy
112-72-1
C14H30O
214.39
MFCD00004757
HLZKNKRTKFSKGZ-UHFFFAOYSA-N
1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks
8209
CHEBI:77417
tetradecan-1-ol
CCCCCCCCCCCCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 37.0°C to 40.0°C
Color White
Boiling Point 289.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)13OH
Quantity 25 g
Beilstein 01, 428
Merck Index 15, 6421
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether, slightly soluble in alcohol
Viscosity 6.4 mPa.s (60°C)
Formula Weight 214.39
Percent Purity 99%
Physical Form Low Melting Solid, Crystalline Powder, Crystals, Flakes, Pellets and/or Chunks
Chemical Name or Material 1-Tetradecanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'