missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-butyl)-5-methyl-1H-pyrazole-3-carbohydrazide, Thermo Scientific™

Catalog Number p-4391244
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
306937-23-5
C9H16N4O
196.254
TWKKUKBETOOWRR-UHFFFAOYSA-N
1-tert-butyl-5-methyl-1h-pyrazole-3-carbohydrazide, maybridge1_008666, 1-tert-butyl-5-methyl-pyrazole-3-carbohydrazide, 1-tert-butyl-5-methylpyrazole-3-carbohydrazide, 1h-pyrazole-3-carboxylicacid, 1-1,1-dimethylethyl-5-methyl-, hydrazide
2735685
1-tert-butyl-5-methylpyrazole-3-carbohydrazide
CC1=CC(=NN1C(C)(C)C)C(=O)NN
This item is not returnable. View return policy
306937-23-5
C9H16N4O
196.254
TWKKUKBETOOWRR-UHFFFAOYSA-N
1-tert-butyl-5-methyl-1h-pyrazole-3-carbohydrazide, maybridge1_008666, 1-tert-butyl-5-methyl-pyrazole-3-carbohydrazide, 1-tert-butyl-5-methylpyrazole-3-carbohydrazide, 1h-pyrazole-3-carboxylicacid, 1-1,1-dimethylethyl-5-methyl-, hydrazide
2735685
1-tert-butyl-5-methylpyrazole-3-carbohydrazide
CC1=CC(=NN1C(C)(C)C)C(=O)NN
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material 1-(tert-Butyl)-5-methyl-1H-pyrazole-3-carbohydrazide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'