missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl 4-(pentafluorophenyl) piperidine-1,4-dicarboxylate, 97%, Thermo Scientific™

Catalog Number p-3586616
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
294885-28-2
C17H18F5NO4
395.33
MFCD09879992
PPKHOXJANSFRFJ-UHFFFAOYSA-N
1-tert-butyl 4-pentafluorophenyl piperidine-1,4-dicarboxylate, 1-tert-butyl 4-2,3,4,5,6-pentafluorophenyl piperidine-1,4-dicarboxylate, o1-tert-butyl o4-2,3,4,5,6-pentakis fluoranyl phenyl piperidine-1,4-dicarboxylate, 1,4-piperidinedicarboxylicacid, 1-1,1-dimethylethyl 4-2,3,4,5,6-pentafluorophenyl ester, piperidine-1,4-dicarboxylic acid o1-tert-butyl ester o4-2,3,4,5,6-pentafluorophenyl ester
11749780
1-O-tert-butyl 4-O-(2,3,4,5,6-pentafluorophenyl) piperidine-1,4-dicarboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
This item is not returnable. View return policy
294885-28-2
C17H18F5NO4
395.33
MFCD09879992
PPKHOXJANSFRFJ-UHFFFAOYSA-N
1-tert-butyl 4-pentafluorophenyl piperidine-1,4-dicarboxylate, 1-tert-butyl 4-2,3,4,5,6-pentafluorophenyl piperidine-1,4-dicarboxylate, o1-tert-butyl o4-2,3,4,5,6-pentakis fluoranyl phenyl piperidine-1,4-dicarboxylate, 1,4-piperidinedicarboxylicacid, 1-1,1-dimethylethyl 4-2,3,4,5,6-pentafluorophenyl ester, piperidine-1,4-dicarboxylic acid o1-tert-butyl ester o4-2,3,4,5,6-pentafluorophenyl ester
11749780
1-O-tert-butyl 4-O-(2,3,4,5,6-pentafluorophenyl) piperidine-1,4-dicarboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 395.32
Percent Purity 95%
Chemical Name or Material 1-tert-Butyl 4-(pentafluorophenyl) piperidine-1, 4-dicarboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'