missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl 4-(9-H-fluoren-9-ylmethyl) hydrogen (2R)-piperazine-1,2,4-tricarboxyl, 97%, Thermo Scientific™

Catalog Number p-3880934
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
209593-18-0
C25H28N2O6
452.507
MFCD02179102
ZVHNNCSUTNWKFC-OAQYLSRUSA-N
r-1-n-boc-4-n-fmoc-2-piperazine carboxylic acid, r-1-n-boc-4-n-fmoc-2-piperazinecarboxylicacid, r-4-9h-fluoren-9-yl methoxy carbonyl-1-tert-butoxycarbonyl piperazine-2-carboxylic acid, 1-boc-4-fmoc-piperazine-2-r-carboxylic acid, 2r-1-tert-butoxycarbonyl-4-9h-fluoren-9-ylmethoxy carbonyl piperazine-2-carboxylic acid, r-piperazine-1,2,4-tricarboxylic acid 1-tert-butyl ester 4-9h-fluoren-9-ylmethyl ester, 1-tert-butyl 4-9-h-fluoren-9-ylmethyl hydrogen 2r-piperazine-1,2,4-tricarboxylate, bestipharma 115-414, r-4-fmoc-1-tert-butoxycarbonyl piperazine-2-carboxylic acid, 2r-4-9h-fluoren-9-ylmethoxycarbonyl-1-2-methylpropan-2-yl oxycarbonyl piperazine-2-carboxylic acid
7128295
(2R)-4-(9H-fluoren-9-ylmethoxycarbonyl)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperazine-2-carboxylic acid
CC(C)(C)OC(=O)N1CCN(CC1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
This item is not returnable. View return policy
209593-18-0
C25H28N2O6
452.507
MFCD02179102
ZVHNNCSUTNWKFC-OAQYLSRUSA-N
r-1-n-boc-4-n-fmoc-2-piperazine carboxylic acid, r-1-n-boc-4-n-fmoc-2-piperazinecarboxylicacid, r-4-9h-fluoren-9-yl methoxy carbonyl-1-tert-butoxycarbonyl piperazine-2-carboxylic acid, 1-boc-4-fmoc-piperazine-2-r-carboxylic acid, 2r-1-tert-butoxycarbonyl-4-9h-fluoren-9-ylmethoxy carbonyl piperazine-2-carboxylic acid, r-piperazine-1,2,4-tricarboxylic acid 1-tert-butyl ester 4-9h-fluoren-9-ylmethyl ester, 1-tert-butyl 4-9-h-fluoren-9-ylmethyl hydrogen 2r-piperazine-1,2,4-tricarboxylate, bestipharma 115-414, r-4-fmoc-1-tert-butoxycarbonyl piperazine-2-carboxylic acid, 2r-4-9h-fluoren-9-ylmethoxycarbonyl-1-2-methylpropan-2-yl oxycarbonyl piperazine-2-carboxylic acid
7128295
(2R)-4-(9H-fluoren-9-ylmethoxycarbonyl)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperazine-2-carboxylic acid
CC(C)(C)OC(=O)N1CCN(CC1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 452.5
Percent Purity 97%
Chemical Name or Material 1-tert-Butyl 4-(9-H-fluoren-9-ylmethyl) hydrogen (2R)-piperazine-1, 2, 4-tricarboxyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'