missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 10 g |
Formula Weight | 182.22 |
Percent Purity | 97% |
Chemical Name or Material | 1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'