Learn More
Thermo Scientific™ 1-Step Heavy Protein IVT Kit
Express stable isotope-labeled (i.e., heavy) proteins by in vitro translation (IVT) for mass spectrometry using these cell-free expression kits.
Brand:  Thermo Scientific™ 88331
Code : PN
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 12122410
  • £807.57 / Each
Estimated Shipment
14-03-2018
Add to basket
Description
Description
The 1-Step Heavy Protein IVT Kit uses a unique HeLa cell lysate supplemented with heavy amino acids for in vitro translation (IVT) of proteins with 90 to 95% isotope incorporation efficiency in less than 8 hours. Heavy proteins expressed using this system can be used as mass spectrometry controls for sample prep loss, digestion efficiency determination or as quantification standards.
Highlights:
  • Efficient – express heavy proteins with 90-95% stable isotope incorporation
  • Functional – uses the human translational machinery to express more biologically active proteins than other IVT systems
  • Flexible – express light proteins or heavy proteins containing 13C615N2 L-lysine and/or 13C615N4 L-arginine
  • Convenient – perform transcription and translation in a single step
  • High performance – greater yields compared to rabbit reticulocyte in vitro translation
Recommended for:
Identify ideal peptides for targeted quantitation; Verify protein digestion efficiency; Control for protein sample prep variability and affinity enrichment loss
Specifications
Specifications
Pierce 1-Step Heavy Protein IVT Kit
Yes
13C6 15N2 L-Lysine, 25uL
40 reactions of 25uL each
Multi-component kit
DNA
40 rxn kit
Videos