missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrenecarboxaldehyde, 98+%, Thermo Scientific Chemicals

Catalog Number p-7045464
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3029-19-4
C17H10O
230.266
MFCD00004139
RCYFOPUXRMOLQM-UHFFFAOYSA-N
1-pyrenecarboxaldehyde, 1-formylpyrene, 1-pyrenealdehyde, pyrenecarboxaldehyde, 3-formylpyrene, 3-pyrenealdehyde, 3-pyrenylaldehyde, pyrene-1-aldehyde, 3-pyrenecarboxaldehyde, 1-pyrenecarbaldehyde
232848
pyrene-1-carbaldehyde
C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C=O
This item is not returnable. View return policy
3029-19-4
C17H10O
230.266
MFCD00004139
RCYFOPUXRMOLQM-UHFFFAOYSA-N
1-pyrenecarboxaldehyde, 1-formylpyrene, 1-pyrenealdehyde, pyrenecarboxaldehyde, 3-formylpyrene, 3-pyrenealdehyde, 3-pyrenylaldehyde, pyrene-1-aldehyde, 3-pyrenecarboxaldehyde, 1-pyrenecarbaldehyde
232848
pyrene-1-carbaldehyde
C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C=O

1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 125°C to 128°C
Quantity 5 g
Beilstein 1874889
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 230.27
Percent Purity 99%
Chemical Name or Material 1-Pyrenecarboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'