missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, for spectroscopy ACS, ACROS Organics™

Brand:  Acros Organics 434360025

Code : 29

Additional Details : CAS Number : 71-23-8 Weight : 3.50000kg Transport : UN number : 1274 Chem class : 3 Pack group : II

Quantity 2.5L
Packaging Glass bottle

 View more versions of this product

Product Code. 10646630

  • £85.90 / 2.50 L


Estimated Shipment
27-06-2018
Add to basket

Description

Description

For UV/VIS spectroscopy
Specifications

Chemical Identifiers

1-Propanol
71-23-8
≥99.5%
CH3CH2CH2OH
Propyl alcohol
For Spectroscopy
For spectroscopy ACS
C3H8O
60.1

Specifications

0.05% max.
For Spectroscopy
0.03% max. (as C2H5CHO)
0.01% max.
For spectroscopy ACS
≥99.5%
C3H8O
Glass bottle
1.3840 to 1.3860
Propyl alcohol
0.0004 meq/g
Clear
− 127.0°C
2.5L
1-Propanol
71-23-8
0.8000g/mL
Authentic
Liquid
0.01% max.
CH3CH2CH2OH
60.1
Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
0.001% max.
0.2% max.
97.0°C
15°C
0.8
Videos
SDS