missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, Spectrophotometric Grade, 99%, Alfa Aesar™

Brand:  Alfa Aesar 022933.M1

Code : NEW

Additional Details : CAS Number : 71-23-8 Weight : 0.01000kg Transport : UN number : 1274 Chem class : 1 Pack group : II

Quantity 4 x 1L

 View more versions of this product

Product Code. 11318217

  • £141.90 / 4 L


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

1-Propanol
Spectrophotometric grade
99%
60.1
71-23-8
Liquid
CH3CH2CH2OH
MFCD00002941

Specifications

1-Propanol
Spectrophotometric grade
Liquid
CH3CH2CH2OH
MFCD00002941
13.3 hPa (10mm Hg) at 20°C
4 x 1L
71-23-8
Clear liquid
99%
60.1
Alcohol-like
Clear
Videos
SDS