missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number p-5547519
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
This item is not returnable. View return policy
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
This item is not returnable. View return policy
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO

Used in making cosmetics, skin and hair preparations, pharmaceuticals, perfumes, lacquer formulations, dye solutions, antifreezes, rubbing alcohols, soaps, window cleaners, acetone and other chemicals and products. Also referred as ropyl alcohol;n-Propan-1-ol;n-Propanol;n-Propyl alcohol, Ethylcarbinol, Optal, Osmosol extra.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 127°C
Color Colorless
Appearance Clear liquid
Density 0.804
Boiling Point 97°C
Quantity 250 mL
Flash Point 15°C (59°F)
Ignition Point 824°C (1515°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.385
Assay Percent Range 99.9%
Vapor Pressure 13.3 hPa (10mm Hg) at 20°C
Beilstein 1098242
Solubility Information Completely soluble in water. Miscible with propylene glycol.
Formula Weight 60.1
Chemical Name or Material 1-Propanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'