missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™

Catalog Number 15635410
Click to view available options
Quantity:
1 L
2.5 L
500 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
This item is not returnable. View return policy
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Melting Point -127°C
Linear Formula CH3CH2CH2OH
UN Number UN1274
Beilstein 1098242
Formula Weight 60.1g/mol
Percent Purity ≥99.5%
Density 0.804g/mL at 25°C
Boiling Point 97°C
Quantity 500 L
Explosion Limit 0.137
Flash Point 22°C (71.6°F)
Ignition Point 700°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.384
Vapor Pressure 14.9mmHg (20°C), 10mmHg (147°C)
Chemical Name or Material 1-Propanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'