missing translation for 'onlineSavingsMsg'
Learn More

1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in dimethylformamide, ACROS Organics™

Brand:  Acros Organics 374661000

Code : 29

Additional Details : CAS Number : 68957-94-8 Weight : 0.15000kg Transport : UN number : 2920 Chem class : 1 Pack group : II


Quantity 100g
Packaging Plastic Bottle

 View more versions of this product

Product Code. 10552563

  • £146.60 / 100g


EU Stock 4
Add to basket


Specifications

Chemical Identifiers

1-Propanephosphonic acid cyclic anhydride
49.0
≥50% solids
318.18
2, 4, 6-Tripropyl-1
68-12-2
51.0
C9H21O6P3
MFCD00006583

Specifications

1-Propanephosphonic acid cyclic anhydride
49.0
1.0900g/mL
≥50% solids
Plastic Bottle
MFCD00006583
1.4500 to 1.4540
2, 4, 6-Tripropyl-1
153.0°C
100g
68-12-2
51.0
Solution
C9H21O6P3
318.18
15, 7978
Solubility in water: miscible.
Clear yellow to brownish
56°C
1.09
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in dimethylformamide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS