missing translation for 'onlineSavingsMsg'
Learn More

1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in dimethylformamide, Thermo Scientific Chemicals

Catalog Number 10240313
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
68957-94-8, 68-12-2
C9H21O6P3
318.18
MFCD00006583
PAQZWJGSJMLPMG-UHFFFAOYSA-N
2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide, 1-propanephosphonic acid cyclic anhydride, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphinane 2,4,6-trioxide, unii-i6egd8839n, 1,3,5,2,4,6-trioxatriphosphorinane, 2,4,6-tripropyl-, 2,4,6-trioxide, propane phosphonic acid anhydride, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane 2,4,6-trioxide, 1-propanephosphonic anhydride in thf, 2,4,6-tri-n-propyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane, 1-propanephosphonic anhydride in toluene, t3p
111923
2,4,6-tripropyl-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide
CCCP1(=O)OP(=O)(OP(=O)(O1)CCC)CCC
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
68957-94-8, 68-12-2
C9H21O6P3
318.18
MFCD00006583
PAQZWJGSJMLPMG-UHFFFAOYSA-N
2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide, 1-propanephosphonic acid cyclic anhydride, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphinane 2,4,6-trioxide, unii-i6egd8839n, 1,3,5,2,4,6-trioxatriphosphorinane, 2,4,6-tripropyl-, 2,4,6-trioxide, propane phosphonic acid anhydride, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane 2,4,6-trioxide, 1-propanephosphonic anhydride in thf, 2,4,6-tri-n-propyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane, 1-propanephosphonic anhydride in toluene, t3p
111923
2,4,6-tripropyl-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide
CCCP1(=O)OP(=O)(OP(=O)(O1)CCC)CCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 49.0
CAS Max % 51.0
Color Brown to Yellow
Density 1.0900g/mL
Boiling Point 153.0°C
Flash Point 56°C
Packaging Plastic Bottle
Refractive Index 1.4500 to 1.4540
Quantity 25 g
Merck Index 15, 7978
Specific Gravity 1.09
Solubility Information Solubility in water: miscible.
Formula Weight 318.18
Percent Purity ≥50% solids
Physical Form Solution
Chemical Name or Material 1-Propanephosphonic acid cyclic anhydride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in dimethylformamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'