missing translation for 'onlineSavingsMsg'
Learn More

1-Piperonylpiperazine, 97%, ACROS Organics™

Brand:  Acros Organics 208930250

Code : 29

Additional Details : CAS Number : 32231-06-4 Weight : 0.07500kg Transport : UN number : 3259 Chem class : 1b Pack group : III


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10702061

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

1-Piperonylpiperazine
32231-06-4
100.0
97%
220.27
97%
96.0
97%
C12H16N2O2
MFCD00005834

Specifications

1-Piperonylpiperazine
32231-06-4
100.0
97%
Authentic
220.27
Glass bottle
White to yellow or yellow-brownish
25g
97%
96.0
Crystalline Solid
97%
C12H16N2O2
MFCD00005834
147.0° to 149.0°C (2.0 mmHg)
38.0° to 42.0°C
>112°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Piperonylpiperazine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS