missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpyrrolidine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7058299
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4096-21-3
C10H13N
147.22
MFCD00015897
VDQQJMHXZCMNMU-UHFFFAOYSA-N
1-phenyl-pyrrolidine, n-phenylpyrrolidine, phenylpyrrolidine, pyrrolidine, 1-phenyl, 1-phenyl-pyrrolidin-3-ol, 1-phenylpyrolidine, n-phenyl pyrrolidine, pubchem9192, acmc-1cttc
77726
1-phenylpyrrolidine
C1CCN(C1)C1=CC=CC=C1
This item is not returnable. View return policy
4096-21-3
C10H13N
147.22
MFCD00015897
VDQQJMHXZCMNMU-UHFFFAOYSA-N
1-phenyl-pyrrolidine, n-phenylpyrrolidine, phenylpyrrolidine, pyrrolidine, 1-phenyl, 1-phenyl-pyrrolidin-3-ol, 1-phenylpyrolidine, n-phenyl pyrrolidine, pubchem9192, acmc-1cttc
77726
1-phenylpyrrolidine
C1CCN(C1)C1=CC=CC=C1

1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents. Store cold 2-8°C.
TRUSTED_SUSTAINABILITY
Boiling Point 133°C to 134°C (19 mmHg)
Refractive Index 1.582
Quantity 5 g
Beilstein 117030
Solubility Information Not miscible in water.
Formula Weight 147.22
Percent Purity ≥98%
Chemical Name or Material 1-Phenylpyrrolidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'